Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B220/CD45R Antibody (HIS24), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143408C
Description
B220/CD45R Monoclonal specifically detects B220/CD45R in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
B220/CD45R | |
Monoclonal | |
DyLight 650 | |
50 mM sodium borate with 0.05% sodium azide | |
PTPRC | |
The immunogen for this antibody was CD45R. | |
0.1 mL | |
Primary | |
Rat | |
IgG2b κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
HIS24 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Mouse | |
Protein A or G purified | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Immunophenotyping, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
5788.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction