Learn More
B220/CD45R Antibody (RM0063-9F14), Alexa Fluor™ 750, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP212168AF750
Catalog No. NP212168A70
Only null left
Add to Cart
Description
Description
B220/CD45R Monoclonal specifically detects B220/CD45R in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
B220/CD45R
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
PTPRC
Mouse leukemia virus-induced pre-B tumor cells
0.1 mL
5788
Store at 4°C in the dark.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
RM0063-9F14
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Rat
Protein A or G purified
Primary
Mouse
IgG2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.