Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B220/CD45R Antibody (YKIX753.22.2), DyLight 650, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10065018C
Description
B220/CD45R Monoclonal specifically detects B220/CD45R in Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
B220/CD45R | |
Monoclonal | |
DyLight 650 | |
50 mM sodium borate with 0.05% sodium azide | |
PTPRC | |
Canine thymocytes | |
0.1 mL | |
Primary | |
This antibody recognizes canine CD45R and was clustered as a canine CD45R at the first canine Leucocyte Antigen workshop. It precipitates an antigen of 220kD from Con A Blasts (1) and may be against a CD45R isoform. It is expressed on subsets of leucocytes (neutrophils are weak) in canine blood of all out bred mongrels and beagles tested. | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
YKIX753.22.2 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Rat | |
Protein G purified | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Immunophenotyping, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
5788.0 | |
Canine | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction