Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B2M Mouse anti-Human, Unconjugated, Clone: 3F9-2C2, Abnova™
Mouse Monoclonal Antibody.
Supplier: Abnova Corporation H00000567M01J
Description
B2M Monoclonal antibody specifically detects B2M in Human samples. It is validated for WB-Ce, IHC-P, IF, S-ELISA, ELISA, WB-Re, WB-Tr, RNAi-Ab, PLA-Ce.Specifications
B2M | |
Monoclonal | |
Unconjugated | |
B2M | |
Mouse | |
100 μg | |
567 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, ELISA, KnockDown, Proximity Ligation Assay | |
3F9-2C2 | |
beta-2-microglobulin | |
BC032589.1 | |
B2M (AAH32589.1, 1 a.a. ∼ 119 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa | |
Primary | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction