Learn More
Brady™ B33 Series Clear, Die-cut Component and Barcode Labels
Catalog No. p-7238948 Shop All Brady Corporation Products
Click to view available options
Labels per Row:
2
5
Width (English):
2.75 in.
3 in.
Clear polyester (B-430) material offers nameplate-like quality when printing labels with barcodes, alphanumeric data, graphic symbols and logos.
Clear polyester (B-430) material offers nameplate-like quality when printing labels with barcodes, alphanumeric data, graphic symbols and logos. A ribbon is required to print this material.
  • Glossy, clear labels are thermal transfer printable
  • Feature a durable permanent acrylic adhesive
  • Excellent resistance to most chemicals, oils and water
  • UL approved, CSA accepted
  • Wide service temperature range
Specifications
Printable Sides Single-side Printable
For Use With (Equipment) Brady Label Printers
Labels per Row 5
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.