Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B3GALNT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109438
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109438 Supplier Invitrogen™ Supplier No. PA5109438
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IERVNWMYFY EYEPIYRQDF HFTLREHSNC SHQNPFLVIL VTSHPSDVKA RQAIRVTWGE KKSWWGYEVL TFFLLGQEAE KEDKMLALSL EDEHLLYGDI IRQDFLDTYN NLTLKTIMAF RWVTEFCPNA KYVMKTDTDV FINTGNLVKY LLNLNHSEKF FTGYPLIDNY SYRGFYQKTH ISYQEYPFKV FPPYCSGLGY IMSRDLVPRI YEMMGHVKPI KFEDVYVGIC LNLLKVNIHI PEDTNLFFLY RIHLDVCQLR RVIAAHGFSS KEIITFWQVM LRNTTCHY.

B3GALNT1 gene ontology annotations related to this gene include protein glycosylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GALNT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene B3GALNT1
Gene Accession No. O75752, Q6AY39, Q920V1
Gene Alias B3galnt1; B3galt3; b3Gal-T3; b3GT3; Beta-1,3-galactosyltransferase 3; beta-1,3-GalNAc-T1; Beta-1,3-GalTase 3; beta-1,3-N-acetylgalactosaminyltransferase 1; beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group); beta-3-GalNAc-T1; beta3GalT3; beta3Gal-T3; beta-3-Gx-T3; Brainiac 1; brainiac1; galactosylgalactosylglucosylceramide beta-D-acetyl-galactosaminyltransferase; galT3; Gb4Cer; GLCT3; GLOB; Globoside blood group; globoside synthase; globotriaosylceramide 3-beta-N-acetylgalactosaminyltransferase; Mbrn 1; Mbrn1; P; P antigen synthase; P blood group globoside; P1; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase 1; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase III; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 3; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 3 (Globoside blood group); UDP-GalNAc:beta-1,3-N-acetylgalactosaminyltransferase 1; UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 1; UDP-GalNAc:betaGlcNAc beta-1,3-galactosaminyltransferase, polypeptide 1 (Globoside blood group); UDP-N-acetylgalactosamine:globotriaosylceramide beta-1,3-N-acetylgalactosaminyltransferase; UNQ531/PRO1074
Gene Symbols B3GALNT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 44-331 of human B3GALNT1 (NP_003772.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26879, 310508, 8706
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.