Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B3GALNT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562231
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562231 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562231 Supplier Invitrogen™ Supplier No. PA562231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WTVETTSFNL LLKTDDDCYI DLEAVFNRIV QKNLDGPNFW WGNFRLNWAV DRTGKWQELE YPSPAYPAFA CGSGYVISKD IVKWLASNSG RLKTYQGEDV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

Beta-1,3-N-acetylgalactosaminyltransferase active in synthesizing a unique carbohydrate structure, GalNAc-beta-1-3GlcNAc, on N- and O-glycans. Has no galactose nor galactosaminyl transferase activity toward any acceptor substrate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GALNT2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene B3GALNT2
Gene Accession No. Q8NCR0
Gene Alias A930105D20Rik; B3GalNAc-T2; B3galnt2; Beta-1,3-GalNAc-T2; beta-1,3-N-acetylgalactosaminyltransferase 2; beta-1,3-N-acetylgalactosaminyltransferase II; C80633; D230016N13Rik; MDDGA11; RGD1306946; UDP-GalNAc:beta-1,3-N-acetylgalactosaminyltransferase 2; UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 2; UDP-GalNAc:betaGlcNAc beta-1,3-galactosaminyltransferase, polypeptide 2
Gene Symbols B3GALNT2
Host Species Rabbit
Immunogen Recombinant protein fragment of Human B3GALNT2 (Product #RP-104089)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148789
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.