Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B3GALNT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113886
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113886 Supplier Invitrogen™ Supplier No. PA5113886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PESFEGTIVW ESQDLHGLVS RNLHKVTVND GGGVLRVITA GEGALPHEFL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

Beta-1,3-N-acetylgalactosaminyltransferase active in synthesizing a unique carbohydrate structure, GalNAc-beta-1-3GlcNAc, on N- and O-glycans. Has no galactose nor galactosaminyl transferase activity toward any acceptor substrate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GALNT2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene B3GALNT2
Gene Accession No. Q8NCR0
Gene Alias A930105D20Rik; B3GalNAc-T2; B3galnt2; Beta-1,3-GalNAc-T2; beta-1,3-N-acetylgalactosaminyltransferase 2; beta-1,3-N-acetylgalactosaminyltransferase II; C80633; D230016N13Rik; MDDGA11; RGD1306946; UDP-GalNAc:beta-1,3-N-acetylgalactosaminyltransferase 2; UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 2; UDP-GalNAc:betaGlcNAc beta-1,3-galactosaminyltransferase, polypeptide 2
Gene Symbols B3GALNT2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human B3GALNT2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148789
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.