Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B3GALNT2 Polyclonal Antibody, Invitrogen™

Catalog No. FEPA5144684
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144684 Supplier Thermo Scientific Supplier No. PA5144684

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Immunogen sequence: WTVETTSFNL LLKTDDDCYI DLEAVFNRIV QKNLDGPNFW WGNFRLNWAV DRTGKWQELE YPSPAYPAFA CGSGYVISKD IVKWLASNSG RLKTYQGEDV Antigen sequence identity: Mouse 94%; Rat 95%

B3GALNT2 Polyclonal antibody specifically detects B3GALNT2 in Human samples. It is validated for Immunocytochemistry, Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GALNT2
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene B3GALNT2
Gene Accession No. Q8NCR0
Gene Alias A930105D20Rik, B3GalNAc-T2, B3galnt2, Beta-1,3-GalNAc-T2, beta-1,3-N-acetylgalactosaminyltransferase 2, beta-1,3-N-acetylgalactosaminyltransferase II, C80633, D230016N13Rik, MDDGA11, RGD1306946, UDP-GalNAc:beta-1,3-N-acetylgalactosaminyltransferase 2, UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 2, UDP-GalNAc:betaGlcNAc beta-1,3-galactosaminyltransferase, polypeptide 2
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 342-441.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148789
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.