Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B3GALT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103650 Supplier Invitrogen™ Supplier No. PA5103650
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total B3GALT2.

This gene is a member of the beta-1,3-galactosyltransferase (beta3GalT) gene family. This family encodes type II membrane-bound glycoproteins with diverse enzymatic functions using different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine). The beta3GalT genes are distantly related to the Drosophila Brainiac gene and have the protein coding sequence contained in a single exon. The beta3GalT proteins also contain conserved sequences not found in the beta4GalT or alpha3GalT proteins. The carbohydrate chains synthesized by these enzymes are designated as type 1, whereas beta4GalT enzymes synthesize type 2 carbohydrate chains. The ratio of type 1:type 2 chains changes during embryogenesis. By sequence similarity, the beta3GalT genes fall into at least two groups: beta3GalT4 and 4 other beta3GalT genes (beta3GalT1-3, beta3GalT5). This gene encodes a protein that functions in N-linked glycoprotein glycosylation and shows strict donor substrate specificity for UDP-galactose.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GALT2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene B3GALT2
Gene Accession No. O43825, O54905
Gene Alias B3galt2; Beta-1,3-galactosyltransferase 2; beta-1,3-GalTase 2; BETA3GALT2; beta-3-galt2; beta3Gal-T2; GLCT2; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase 2; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 2; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase-II; UDP-galactose:2-acetamido-2-deoxy-D-glucose 3beta-galactosyltransferase 2; Unknown (protein for MGC:127611); zgc:76904; zgc:76904 protein
Gene Symbols B3GALT2
Host Species Rabbit
Immunogen A synthesized peptide derived from human B3GALT2(Accession O43825), corresponding to amino acid residues S388-H422.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26878, 8707
Target Species Human, Mouse, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.