Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B3GALT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543534
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543534 Supplier Invitrogen™ Supplier No. PA543534
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EREQARHGDL LLLPALRDAY ENLTAKVLAM LAWLDEHVAF EFVLKADDDS Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 77%.

B3GALT6, Beta-1, 3-galactosyltransferase that transfers galactose from UDP-galactose to substrates with a terminal beta-linked galactose residue. B3GALT6 has a preference for galactose-beta-1, 4-xylose that is found in the linker region of glycosaminoglycans, such as heparan sulfate and chondroitin sulfate. B3GALT6 has no activity towards substrates with terminal glucosamine or galactosamine residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GALT6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene B3GALT6
Gene Accession No. Q96L58
Gene Alias b3galt6; BB129894; Beta-1,3-galactosyltransferase 6; beta-1,3-GalTase 6; beta3GalT6; beta3Gal-T6; EDSP2; GAG GalTII; galactosyltransferase II; Galactosylxylosylprotein 3-beta-galactosyltransferase; GalTII; SEMDJL1; UDP-Gal:betaGal beta 1,3-galactosyltransferase 6; UDP-Gal:betaGal beta 1,3-galactosyltransferase polypeptide 6; UDP-Gal:betaGal beta 1,3-galactosyltransferase, polypeptide 6; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 6
Gene Symbols B3GALT6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human B3GALT6 (aa 110-159).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126792
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.