Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B3GALT6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735951 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5951 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5951 Supplier Proteintech Group Inc Supplier No. 550491AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody is specific to B3GALT6.

B3GALT6, Beta-1, 3-galactosyltransferase that transfers galactose from UDP-galactose to substrates with a terminal beta-linked galactose residue. B3GALT6 has a preference for galactose-beta-1, 4-xylose that is found in the linker region of glycosaminoglycans, such as heparan sulfate and chondroitin sulfate. B3GALT6 has no activity towards substrates with terminal glucosamine or galactosamine residues.

Specifications

Antigen B3GALT6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene B3GALT6
Gene Accession No. Q91Z92, Q96L58
Gene Alias B3GALT6, Beta 1,3 GalTase 6, Beta3Gal T6, beta3GalT6, GAG GalTII, Galactosyltransferase II
Gene Symbols B3galt6
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117592, 126792
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.