Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B3GALTL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101457
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101457 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101457 Supplier Invitrogen™ Supplier No. PA5101457
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total B3GALTL.

The protein encoded by this gene is a beta-1, 3-glucosyltransferase that transfers glucose to O-linked fucosylglycans on thrombospondin type-1 repeats (TSRs) of several proteins. The encoded protein is a type II membrane protein. Defects in this gene are a cause of Peters-plus syndrome (PPS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GALTL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene B3GLCT
Gene Accession No. Q6Y288, Q8BHT6
Gene Alias B3galtl; B3GLCT; B3Glc-T; B3GTL; beta 1,3-galactosyltransferase-like; beta 3-glucosyltransferase; beta 3-glycosyltransferase-like; beta-1,3-glucosyltransferase; Beta3Glc-T; beta-3-glucosyltransferase; beta-3-glycosyltransferase-like; Gal-T; Gm1057; UDP-GAL:beta-GlcNAc beta-1,3-galactosyltransferase-like
Gene Symbols B3GLCT
Host Species Rabbit
Immunogen A synthesized peptide derived from human B3GLCT(Accession Q6Y288), corresponding to amino acid residues S461-L498.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145173, 381694
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.