Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B3GALTL Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549563
Description
The protein encoded by this gene is a beta-1,3-glucosyltransferase that transfers glucose to O-linked fucosylglycans on thrombospondin type-1 repeats (TSRs) of several proteins. The encoded protein is a type II membrane protein. Defects in this gene are a cause of Peters-plus syndrome (PPS).
Specifications
B3GALTL | |
Polyclonal | |
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6Y288 | |
B3GLCT | |
synthetic peptide corresponding to 470-496aa of human B3GLT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
B3GLCT | |
Beta-1, 3-glucosyltransferase, Beta3Glc-T, 241-, Beta-3-glycosyltransferase-like, B3GALTL, B3GTL | |
Rabbit | |
Affinity chromatography | |
RUO | |
145173 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction