Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B3GAT1 Mouse anti-Human, Clone: SPM129, Abnova™

Catalog No. 89933615 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-615 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-615 Supplier Abnova Corporation Supplier No. MAB13030
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native human B3GAT1.

The protein encoded by this gene is a member of the glucuronyltransferase gene family. These enzymes exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product functions as the key enzyme in a glucuronyl transfer reaction during the biosynthesis of the carbohydrate epitope HNK-1 (human natural killer-1, also known as CD57 and LEU7). Alternate transcriptional splice variants have been characterized. [provided by RefSeq]

Specifications

Antigen B3GAT1
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SPM129
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native human B3GAT1.
Dilution Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Alias CD57/GLCATP/GlcAT-P/GlcUAT-P/HNK-1/HNK1/LEU7/NK-1
Gene Symbols B3GAT1
Host Species Mouse
Immunogen Human peripheral blood mononuclear cells.
Purification Method Polyethylene Glycol (PEG) precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27087
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.