Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B3GAT1 Mouse anti-Human, Unconjugated, Clone: HNK-1 + NK-1, Abnova™

Catalog No. 89337695 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-695 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-695 Supplier Abnova Corporation Supplier No. MAB21202
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

B3GAT1 Monoclonal antibody specifically detects B3GAT1 in Human samples. It is validated for IHC-P, IF.

Specifications

Antigen B3GAT1
Applications Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone HNK-1 + NK-1
Conjugate Unconjugated
Description beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P)
Gene B3GAT1
Gene Alias CD57, GLCATP, GlcAT-P, GlcUAT-P, HNK-1, HNK1, LEU7, NK-1
Host Species Mouse
Immunogen Human peripheral blood mononuclear cells
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 27087
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.