Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B3GAT1, Rabbit anti-Human, Clone: E20-I, Abnova™

Catalog No. 89998695 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-695 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-695 Supplier Abnova Corporation Supplier No. MAB15988

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

Specifications

Antigen B3GAT1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone E20-I
Conjugate Unconjugated
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) The optimal working dilution should be determined by the end user.
Formulation In 20 mM Tris-HCl buffer, pH 8.0 (20 mg/mL BSA, 0.05% Sodium Azide).
Gene beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P)
Gene Alias CD57/GLCATP/GlcAT-P/GlcUAT-P/HNK-1/HNK1/LEU7/NK-1
Gene Symbols B3GAT1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human B3GAT1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27087
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.