Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ B3GAT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562383
Description
Immunogen sequence: AASGLLFTHL VVLTPKAQRL REGEPGWVHP RGVEQRNKAL DWLRGRGGAV GGEKDPPPPG TQGVVY Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
The protein encoded by this gene is a member of the glucuronyltransferase gene family, enzymes that exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product catalyzes the formation of the glycosaminoglycan-protein linkage by way of a glucuronyl transfer reaction in the final step of the biosynthesis of the linkage region of proteoglycans.
Specifications
B3GAT3 | |
Polyclonal | |
Unconjugated | |
B3GAT3 | |
2810405M13Rik; 3-beta-glucuronosyltransferase; B3GAT3; Beta-1,3-glucuronyltransferase 3; beta-1,3-glucuronyltransferase 3 (glucuronosyltransferase I); galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 3; GLCATI; GlcAT-I; glcUAT-I; glucuronosyltransferase I; glucuronosyltransferase-I; glucuronyltransferase I; JDSCD; LOW QUALITY PROTEIN: galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 3; Sqv-8-like protein; UDP-GlcUA:Gal beta-1,3-Gal-R glucuronyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26229 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94766 | |
B3GAT3 | |
Recombinant protein corresponding to Human B3GAT3. Recombinant protein control fragment (Product #RP-101363). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction