Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B3GAT3 Polyclonal Antibody

Catalog No. PIPA542943
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542943 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542943 Supplier Invitrogen™ Supplier No. PA542943

Rabbit Polyclonal Antibody

Peptide sequence: PPLRAAAEQL RQKDLRISQL QAELRRPPPA PAQPPEPEAL PTIYVVTPTY Sequence homology: Cow: 100%; Dog: 100%; Goat: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

The protein encoded by this gene is a member of the glucuronyltransferase gene family, enzymes that exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product catalyzes the formation of the glycosaminoglycan-protein linkage by way of a glucuronyl transfer reaction in the final step of the biosynthesis of the linkage region of proteoglycans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GAT3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene B3GAT3
Gene Accession No. O94766
Gene Alias 2810405M13Rik; 3-beta-glucuronosyltransferase; B3GAT3; Beta-1,3-glucuronyltransferase 3; beta-1,3-glucuronyltransferase 3 (glucuronosyltransferase I); galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 3; GLCATI; GlcAT-I; glcUAT-I; glucuronosyltransferase I; glucuronosyltransferase-I; glucuronyltransferase I; JDSCD; LOW QUALITY PROTEIN: galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 3; Sqv-8-like protein; UDP-GlcUA:Gal beta-1,3-Gal-R glucuronyltransferase
Gene Symbols B3GAT3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human B3GAT3 (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26229
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.