Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ B3GNT1 Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547860
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) B4GalT1 and rhB3GNT 2 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
Beta-1,4-glucuronyltransferase involved in O- mannosylation of alpha-dystroglycan (DAG1). Transfers a glucuronic acid (GlcA) residue onto a xylose (Xyl) acceptor to produce the glucuronyl-beta-1,4-xylose-beta disaccharide primer, which is further elongated by LARGE, during synthesis of phosphorylated O- mannosyl glycan (PubMed:25279699, PubMed:25279697). Phosphorylated O-mannosyl glycan is a carbohydrate is a carbohydrate structure present in alpha-dystroglycan (DAG1), which is required for binding laminin G-like domain-containing extracellular proteins with high affinity (PubMed:25279699, PubMed:25279697). Required for axon guidance; via its function in O-mannosylation of alpha- dystroglycan (DAG1).
Specifications
| B3GNT1 | |
| Polyclonal | |
| Unconjugated | |
| B4GAT1 | |
| 1500032M01Rik; B3GNT1; B3GN-T1; B3GNT1 (HGNC:15685); B3gnt6; B4GAT1; beta-1,3-N-acetylglucosaminyltransferase bGnT-6; beta-1,4-glucuronyltransferase 1; BETA3GNT1; BETA3GNTI; EC 2.4.1.149; I-beta-1,3-N-acetylglucosaminyltransferase; iGAT; iGNT; MDDGA13; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase; Poly-N-acetyllactosamine extension enzyme; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 | |
| Sheep | |
| Antigen Affinity Chromatography | |
| RUO | |
| 108902, 11041 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O43505, Q8BWP8 | |
| B4GAT1 | |
| CHO-derived recombinant human beta-1,3-N-acetylglucosaminyltransferase 1/B3GNT1 Asp43-Cys415. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction