Learn More
Invitrogen™ B3GNT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552237
Description
Immunogen sequence: DTEFVFKGDD DVFVNTHHIL NYLNSLSKTK AKDLFIGDVI HNAGPHRDKK LKYYIPEVVY SGLYPPYAGG GGFLYSGHLA LRLYHITDQV HLYPIDDVYT GMCLQKLGLV PEKHKGFRTF DIEEKNKNNI CSYVDLMLVH SRKPQEMI Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
This gene encodes a member of the beta-1,3-N-acetylglucosaminyltransferase family. This enzyme is a type II transmembrane protein. It prefers the substrate of lacto-N-neotetraose, and is involved in the biosynthesis of poly-N-acetyllactosamine chains.
Specifications
B3GNT2 | |
Polyclonal | |
Unconjugated | |
B3GNT2 | |
3-Gn-T1; 3-Gn-T2; 3-N-a; B3GALT7; B3GNT; B3GNT1; B3GN-T1; B3GNT2; B3GN-T2; B3GNT-2; beta-1; Beta-1,3-galactosyltransferase 7; Beta-1,3-GalTase 7; beta-1,3-Gn-T1; beta-1,3-Gn-T2; Beta-1,3-N-acetylglucosaminyltransferase 1; beta-1,3-N-acetylglucosaminyltransferase bGnT-1; beta-1,3-N-acetylglucosaminyltransferase bGnT-2; BETA3GNT; beta3Gn-T1; beta3Gn-T2; Beta-3-Gx-T7; BGnT-1; BGNT2; BGnT-2; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 2; UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 7; UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 7; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10678 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NY97 | |
B3GNT2 | |
Recombinant protein corresponding to Human B3GNT2. Recombinant protein control fragment (Product #RP-102031). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.