Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B3GNT3 Monoclonal Antibody (1A2)
GREENER_CHOICE

Catalog No. PIMA521011
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521011 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521011 Supplier Thermo Scientific Supplier No. MA521011
Add to Cart
Add to Cart

B3GNT3 Monoclonal Antibody (1A2)

B3GNT3 Monoclonal Antibody for ICC/IF, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GNT3
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1A2
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene B3gnt3
Gene Accession No. Q9Y2A9
Gene Alias 2210008L19Rik; B3GALT8; B3GAL-T8; B3gnt3; B3GN-T3; B3GNT-3; Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase; Beta-1,3-galactosyltransferase 8; Beta-1,3-GalTase 8; beta-1,3-Gn-T3; beta-1,3-N-acetylglucosaminyltransferase 3; beta-1,3-N-acetylglucosaminyltransferase bGnT-3; beta3GalT8; beta3Gn-T3; beta-3-Gx-T8; BGnT-3; core 1 beta-1,3-N-acetylglucosaminyltransferase; core 1 extending beta-1,3-N-acetylglucosaminyltransferase; Core1-beta3GlcNAcT; HP10328; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 3; putative type II membrane protein; TMEM3; Transmembrane protein 3; UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 8; UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 8; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3; UNQ637/PRO1266
Gene Symbols B3gnt3
Host Species Mouse
Immunogen B3GNT3 (NP_055071, 135 a.a. ~ 208 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10331
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.