Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ B3GNT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555045
Description
Immunogen sequence: LLQDVPPSKC AQPVFLLLVI KSSPSNYVRR ELLRRTWGRE RKVRGLQLRL LFLVGTASNP HEARKVNRLL ELEAQTHGDI LQWDFHDSFF NLTLKQVLFL QWQETRCANA SFVLNGDDDV FAHTDNMVFY LQDHDPGRHL FV Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 73%.
This gene encodes a member of the beta-1, 3-N-acetylglucosaminyltransferase family. This enzyme is a type II transmembrane protein and contains a signal anchor that is not cleaved. It prefers the substrates of lacto-N-tetraose and lacto-N-neotetraose, and is involved in the biosynthesis of poly-N-acetyllactosamine chains and the biosynthesis of the backbone structure of dimeric sialyl Lewis a. It plays dominant roles in L-selectin ligand biosynthesis, lymphocyte homing and lymphocyte trafficking.
Specifications
B3GNT3 | |
Polyclonal | |
Unconjugated | |
B3gnt3 | |
2210008L19Rik; B3GALT8; B3GAL-T8; B3gnt3; B3GN-T3; B3GNT-3; Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase; Beta-1,3-galactosyltransferase 8; Beta-1,3-GalTase 8; beta-1,3-Gn-T3; beta-1,3-N-acetylglucosaminyltransferase 3; beta-1,3-N-acetylglucosaminyltransferase bGnT-3; beta3GalT8; beta3Gn-T3; beta-3-Gx-T8; BGnT-3; core 1 beta-1,3-N-acetylglucosaminyltransferase; core 1 extending beta-1,3-N-acetylglucosaminyltransferase; Core1-beta3GlcNAcT; HP10328; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 3; putative type II membrane protein; TMEM3; Transmembrane protein 3; UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 8; UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 8; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3; UNQ637/PRO1266 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10331 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.12 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2A9 | |
B3gnt3 | |
Recombinant protein corresponding to Human B3GNT3. Recombinant protein control fragment (Product #RP-88799). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction