Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B3GNT3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | B3GNT3 |
---|---|
Concentration | 0.74 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1984
![]() ![]() |
Proteintech Group Inc
180981AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a member of the beta-1, 3-N-acetylglucosaminyltransferase family. This enzyme is a type II transmembrane protein and contains a signal anchor that is not cleaved. It prefers the substrates of lacto-N-tetraose and lacto-N-neotetraose, and is involved in the biosynthesis of poly-N-acetyllactosamine chains and the biosynthesis of the backbone structure of dimeric sialyl Lewis a. It plays domit roles in L-selectin ligand biosynthesis, lymphocyte homing and lymphocyte trafficking.Specifications
B3GNT3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q5JCS9, Q9Y2A9 | |
10331, 72297 | |
B3GNT3 Fusion Protein Ag12061 | |
Primary | |
-20°C | |
B3GNT3 |
0.74 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B3GAL T8, B3GALT8, B3GN T3, B3GNT 3, B3GNT3, Beta 1,3 GalTase 8, Beta 1,3 Gn T3, Beta 3 Gx T8, Beta3Gal T8, Beta3GalT8, beta3Gn T3, BGnT 3, Core1 beta3GlcNAcT, HP10328, TMEM3 | |
B3gnt3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title