Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B3GNT4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590801
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590801 Supplier Invitrogen™ Supplier No. PA590801
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APPAQLLAYE SREFDDILQW DFTEDFFNLT LKELHLQRWV VAACPQAHFM LKGDDDVFVH VPNVLEFLDG WDPAQDLLVG DVIRQALPNR NTKVKYFIPP SMYRATHYPP YAGGGGYVMS RATVRRLQAI MEDAELFPID D; Positive Samples: Rat brain, Rat intestine; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.

This gene encodes a member of the beta-1,3-N-acetylglucosaminyltransferase protein family. The encoded enzyme is involved in the biosynthesis of poly-N-acetyllactosamine chains and prefers lacto-N-neotetraose as a substrate. It is a type II transmembrane protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GNT4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene B3GNT4
Gene Accession No. Q1RLK6, Q9C0J1
Gene Alias 1010001G17Rik; B3GNT4; B3GN-T4; beta-1,3-Gn-T4; beta-1,3-N-acetylglucosaminyltransferase 4; beta-1,3-N-acetylglucosaminyltransferase bGn-T4; beta3Gn-T4; BGnT-4; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 4; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4; UNQ1898/PRO4344
Gene Symbols B3GNT4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 165-305 of human B3GNT4 (NP_1103921).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231727, 288752, 79369
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.