Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B3GNT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553578
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553578 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553578 Supplier Invitrogen™ Supplier No. PA553578
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GAPPIRDKSS KYYVSYEMYQ WPAYPDYTAG AAYVISGDVA AKVYEASQTL NSSLYIDDVF MGLCANKIGI VPQDHVFFSG EGKTPYHPCI YEKMMTSHGH LEDLQDLWKN ATDPKV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.

B3GNT5 is a member of the beta-1,3-N-acetylglucosaminyltransferase family. This enzyme is a type II membrane protein. It exhibits strong activity to transfer GlcNAc to glycolipid substrates and is identified as the most likely candidate for lactotriaosylceramide synthase. This enzyme is essential for the expression of Lewis X epitopes on glycolipids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GNT5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene B3GNT5
Gene Accession No. Q9BYG0
Gene Alias B3gnt5; B3GN-T5; beta 1,3 N-acetyglucosaminyltransferase Lc3 synthase; beta-1,3-Gn-T5; beta-1,3-N-acetylglucosaminyltransferase 5; beta-1,3-N-acetylglucosaminyltransferase bGnT-5; beta-1,3-N-acetylglucosaminyltransferase-5; beta3GnT5; beta3Gn-T5; BGnT-5; lactosylceramide 1,3-N-acetyl-beta-D-glucosaminyltransferase; Lactotriaosylceramide synthase; lc(3)Cer synthase; lc3 synthase; UDP-GlcNAc:beta-Gal beta-1,3-N-acetylglucosaminyltransferase 5; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5
Gene Symbols B3GNT5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human B3GNT5. Recombinant protein control fragment (Product #RP-90486).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84002
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.