Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ B3GNT6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558936
Description
Immunogen sequence: QQWFLQAPRS PREERSPQEE TPEGPTDA Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 43%.
The protein encoded by this gene is a beta-1,3-N-acetylglucosaminyltransferase that adds an N-acetylglucosamine moiety to N-acetylgalactosamine-modified serine or threonine. The encoded enzyme is responsible for creating the core 3 structure of O-glycans, which are important components of mucin-type glycoproteins.
Specifications
B3GNT6 | |
Polyclonal | |
Unconjugated | |
B3GNT6 | |
acetylgalactosaminyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase; B3GNT6; B3Gn-T6; BC039789; beta-1,3-Gn-T6; beta-1,3-N-acetylglucosaminyltransferase 6; beta-1,3-N-acetylglucosaminyltransferase protein; beta3Gn-T6; BGnT-6; core 3 synthase; Core3; Core-3; RGD1310926; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 (core 3 synthase) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192134 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZMB0 | |
B3GNT6 | |
Recombinant protein corresponding to Human B3GNT6. Recombinant protein control fragment (Product #RP-104289). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction