Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B3GNT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590159
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590159 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590159 Supplier Invitrogen™ Supplier No. PA590159
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLQAPRSPRE ERSPQEETPE GPTDAPAADE PPSELVPGPP CVANASANAT ADFEQLPARI QDFLRYRHCR HFPLLWDAPA KCAGGRGVFL LLAVKSAPEH YERRELIRRT WGQERSYGGR PVRRLFLLGT PGPEDEARAE RLAELVALEA REHGDVLQWA FADTFLNLTL K; Positive Samples: Rat stomach; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.

The protein encoded by this gene is a beta-1,3-N-acetylglucosaminyltransferase that adds an N-acetylglucosamine moiety to N-acetylgalactosamine-modified serine or threonine. The encoded enzyme is responsible for creating the core 3 structure of O-glycans, which are important components of mucin-type glycoproteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GNT6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene B3GNT6
Gene Alias acetylgalactosaminyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase; B3GNT6; B3Gn-T6; BC039789; beta-1,3-Gn-T6; beta-1,3-N-acetylglucosaminyltransferase 6; beta-1,3-N-acetylglucosaminyltransferase protein; beta3Gn-T6; BGnT-6; core 3 synthase; Core3; Core-3; RGD1310926; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 (core 3 synthase)
Gene Symbols B3GNT6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-200 of human B3GNT6 (NP_6196513).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292325
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.