Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B3GNT6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | B3GNT6 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3353
![]() ![]() |
Proteintech Group Inc
212911AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Specifications
B3GNT6 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q6ZMB0 | |
192134, 272411 | |
B3GNT6 Fusion Protein Ag15825 | |
Primary | |
-20°C | |
B3GNT6 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B3Gn T6, B3GNT6, Beta 1,3 Gn T6, Beta3Gn T6, BGnT 6, Core 3 synthase, IMAGE:4907098 | |
B3gnt6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title