Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B3GNT8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560459
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560459 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560459 Supplier Invitrogen™ Supplier No. PA560459
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TPANPEPTLP ANLSTRLGQT IPLPFAYWNQ QQWRLGSLPS GDSTETGGCQ AWGAAAATEI PDFASYPKDL RRFLLSAACR SFPQWLP Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 69%.

B3GNT8 is a galactosyltransferase involved in the synthesis of poly-N-acetyllactosamine (polyLacNAc), a linear chain of repeating LacNAc units made up of galactose (Gal) and N-acetylglucosamine (GlcNAc) with the structure (Gal-beta-1-4-GlcNAc-beta-1-3)n. By genomic sequence analysis, the B3GNT8 gene is mapped to chromosome 19q13.2. It was showed that a soluble form of B3GNT8 overexpressed by transfected HEK293 cells selectively transferred GlcNAc from UDP-GlcNAc to the nonreducing terminus of Gal-beta-1-4-GlcNAc-alpha-p-nitrophenyl phosphate and to lactoside-alpha-benzoyl. It did not utilize keratan sulfates or polylactosamine oligosaccharide as substrate. B3GNT8 activity required Mn (2+) and showed less efficiency with Co (2+). The pH optimum was between 7 and 7.5. B3GNT8 also transferred GlcNAc onto alpha-1-acid glycoprotein and ovomucoid, which possess tetraantennary complex type and pentaantennary complex type N-glycans. With a tetraantennary N-glycan substrate, B3GNT8 appeared to prefer the beta-1-2 branch over the beta-1-6 branch. When overexpressed in HCT15 human colon cancer cells, B3GNT8 increased cell surface expression of both polyLacNAc and beta-1-6-branched N-glycans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GNT8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene B3GNT8
Gene Accession No. Q7Z7M8
Gene Alias B3GALT7; B3GNT8; beta galactosyltransferase BGALT15; beta-1,3-Gn-T8; beta-1,3-N-acetylglucosaminyltransferase 8; beta1,3-N-acetylglucosaminyltransferase 8; beta3Gn-T8; BGALT15; BGnT-8; UDP-Gal:betaGal beta 1,3-galactosyltransferase polypeptide 7; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 8
Gene Symbols B3GNT8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human B3GNT8. Recombinant protein control fragment (Product #RP-99175).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374907
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.