Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B3GNT9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566526
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566526 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566526 Supplier Invitrogen™ Supplier No. PA566526
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VFLGMCLQRLR LTPEPHPAFR TFGIPQPSAA PHLSTFD Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.

B3GNT9 gene ontology annotations related to this gene include poly-N-acetyllactosamine biosynthetic process; protein glycosylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GNT9
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene B3GNT9
Gene Accession No. Q6UX72
Gene Alias B3GNT9; beta-1,3-galactosyltransferase-related protein; beta-1,3-Gn-T9; beta-1,3-N-acetylglucosaminyltransferase 9; beta3Gn-T9; BGnT-9; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 9; UNQ1922/PRO4397
Gene Symbols B3GNT9
Host Species Rabbit
Immunogen Recombinant Human B3GNT9. Recombinant protein control fragment (Product #RP-107191).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84752
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.