Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B4GALNT1 Monoclonal Antibody (4G11B1)
GREENER_CHOICE

Catalog No. FERMA548465
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548465 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548465 Supplier Invitrogen™ Supplier No. MA548465
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B4GALNT1 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

GM2 and GD2 gangliosides are sialic acid-containing glycosphingolipids. GalNAc-T is the enzyme involved in the biosynthesis of G(M2) and G(D2) glycosphingolipids. GalNAc-T catalyzes the transfer of GalNAc into G(M3) and G(D3) by a beta-1,4 linkage, resulting in the synthesis of G(M2) and G(D2), respectively. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B4GALNT1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone 4G11B1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene B4GALNT1
Gene Accession No. Q00973, Q09200
Gene Alias (N-acetylneuraminyl)-galactosylglucosylceramide; 4933429D13Rik; B4galnt1; Beta-1,4 N-acetylgalactosaminyltransferase 1; beta1,4GalNAC-T; beta-1,4-N-acetyl-galactosaminyl transferase 1; beta-1,4-N-acetyl-galactosaminyltransferase 1; beta-4N-acetylgalactosaminyltransferase; GALGT; Galgt1; GalNAcT; Gal-NAc-T; GalNAc-T; GD2 synthase, GM2 synthase; Ggm2; Ggm-2; GM2/GD2 synthase; SIAT2; SPG26; UDP-Gal:betaGlcNAc beta-1,4-N-acetylgalactosaminyltransferase transferase 1; UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramide N-acetylgalactosaminyltransferase (GalNAc-T); UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramide-beta-1, 4-N-acetylgalactosaminyltransferase
Gene Symbols B4GALNT1
Host Species Mouse
Immunogen Purified recombinant fragment of human B4GALNT1 (AA: 26-225) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14421, 2583
Target Species Human, Mouse, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.