Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B4GALNT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101460
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101460 Supplier Invitrogen™ Supplier No. PA5101460
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total B4GALNT1.

GM2 and GD2 gangliosides are sialic acid-containing glycosphingolipids. GalNAc-T is the enzyme involved in the biosynthesis of G(M2) and G(D2) glycosphingolipids. GalNAc-T catalyzes the transfer of GalNAc into G(M3) and G(D3) by a beta-1,4 linkage, resulting in the synthesis of G(M2) and G(D2), respectively. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B4GALNT1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene B4GALNT1
Gene Accession No. Q00973, Q09200, Q10468
Gene Alias (N-acetylneuraminyl)-galactosylglucosylceramide; 4933429D13Rik; B4galnt1; Beta-1,4 N-acetylgalactosaminyltransferase 1; beta1,4GalNAC-T; beta-1,4-N-acetyl-galactosaminyl transferase 1; beta-1,4-N-acetyl-galactosaminyltransferase 1; beta-4N-acetylgalactosaminyltransferase; GALGT; Galgt1; GalNAcT; Gal-NAc-T; GalNAc-T; GD2 synthase, GM2 synthase; Ggm2; Ggm-2; GM2/GD2 synthase; SIAT2; SPG26; UDP-Gal:betaGlcNAc beta-1,4-N-acetylgalactosaminyltransferase transferase 1; UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramide N-acetylgalactosaminyltransferase (GalNAc-T); UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramide-beta-1, 4-N-acetylgalactosaminyltransferase
Gene Symbols B4GALNT1
Host Species Rabbit
Immunogen A synthesized peptide derived from human B4GALNT1(Accession Q00973), corresponding to amino acid residues D290-R340.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14421, 2583, 64828
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.