Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B4GALNT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553282
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553282 Supplier Invitrogen™ Supplier No. PA553282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVTGVTLTGE GQADLTLVSP GLDQLNRQLQ LVTYSSRSYQ TNTADTVRFS TEGHEAAFTI RIRHPPNPRL YPPGSLPQGA QYNISALVTI ATKTFLRYDR LRALITSIRR FYPTVTVVIA DDSDKPERVS GPYV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.

GM2 and GD2 gangliosides are sialic acid-containing glycosphingolipids. GalNAc-T is the enzyme involved in the biosynthesis of G(M2) and G(D2) glycosphingolipids. GalNAc-T catalyzes the transfer of GalNAc into G(M3) and G(D3) by a beta-1,4 linkage, resulting in the synthesis of G(M2) and G(D2), respectively. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B4GALNT1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene B4GALNT1
Gene Accession No. Q00973
Gene Alias (N-acetylneuraminyl)-galactosylglucosylceramide; 4933429D13Rik; B4galnt1; Beta-1,4 N-acetylgalactosaminyltransferase 1; beta1,4GalNAC-T; beta-1,4-N-acetyl-galactosaminyl transferase 1; beta-1,4-N-acetyl-galactosaminyltransferase 1; beta-4N-acetylgalactosaminyltransferase; GALGT; Galgt1; GalNAcT; Gal-NAc-T; GalNAc-T; GD2 synthase, GM2 synthase; Ggm2; Ggm-2; GM2/GD2 synthase; SIAT2; SPG26; UDP-Gal:betaGlcNAc beta-1,4-N-acetylgalactosaminyltransferase transferase 1; UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramide N-acetylgalactosaminyltransferase (GalNAc-T); UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosylglucosylceramide-beta-1, 4-N-acetylgalactosaminyltransferase
Gene Symbols B4GALNT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human B4GALNT1. Recombinant protein control fragment (Product #RP-90252).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2583
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.