Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B4GALNT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NBP191229
Description
B4GALNT2 Polyclonal specifically detects B4GALNT2 in Human, Mouse, Rhesus Macaque samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
B4GALNT2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200, Immunohistochemistry-Frozen | |
4 GalNAcT/Sda-GalNAcT, B4GALT, beta 1,4 N-acetylgalactosaminyltransferase/betal, beta-1,4 N-acetylgalactosaminyltransferase 2, beta-1,4-N-acetyl-galactosaminyl transferase 2, beta-4-N-acetylgalactosaminyltransferase, GALGT2, MGC142235, MGC142237, sd(a) beta-1,4-GalNAc transferase, sda beta-1,4-GalNAc transferase, UDP-GalNAc:Neu5Aca2-3Galb-R b1,4-N-acetylgalactosaminyltransferase | |
Rabbit | |
Affinity Purified | |
RUO | |
124872 | |
Human, Mouse, Rhesus Macaque | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
B4GALNT2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LLPEERLRNLFSYDGIWLFPKNQCKCEANKEQGGYNFQDAYGQSDLPAVKARRQAEFEHFQRREGLPRPLPLLVQPNLPFGYPVHGVEVMPLHTVPIPGLQFEGPDA | |
0.1 mL | |
Primary | |
Specificity of human B4GALNT2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction