Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ B4GALNT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570775
Description
This target displays homology in the following species: Human: 100%.
B4GALNT2 catalyzes the last step in the biosynthesis of the human Sd(a) antigen through the addition of an N-acetylgalactosamine residue via a beta-1,4 linkage to a subterminal galactose residue substituted with an alpha-2,3-linked sialic acid. B4GALNT2 also catalyzes the last step in the biosynthesis of the Cad antigen (Montiel et al., 2003 [PubMed 12678917]).
Specifications
B4GALNT2 | |
Polyclonal | |
Unconjugated | |
B4GALNT2 | |
AI593864; B4galnt2; B4GALT; beta 1,4 N-acetylgalactosaminyltransferase/betal,4 GalNAcT/Sda-GalNAcT; beta-1,4 N-acetylgalactosaminyltransferase 2; beta-1,4-N-acetyl-galactosaminyl transferase 2; beta-1,4-N-acetyl-galactosaminyltransferase 2; beta-4-N-acetylgalactosaminyltransferase; CT-Gal NAc transferase; Dlb1; Dlb-1; dolichos lectin binding 1; Galg2; Galgt2; Ggm3; Igf2bp1; N-acetylgalactosaminyltransferase; sd(a) beta-1,4-GalNAc transferase; sda beta-1,4-GalNAc transferase; UDP-GalNAc:Neu5Aca2-3Galb-R b1,4-N-acetylgalactosaminyltransferase; UDP-GalNAc:Neu5Acalpha2-3Galbeta-R beta1,4-N-acetylgalactosaminyltransferase; UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosyl-N-acetylglucosaminylpolypeptide-beta-1, 4-N-acetylgalactosaminyltransferase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
124872 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NHY0 | |
B4GALNT2 | |
synthetic peptide directed towards the N-terminal region of human B4GALNT2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction