Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B4GALNT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103653
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103653 Supplier Invitrogen™ Supplier No. PA5103653
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total B4GALNT2.

B4GALNT2 catalyzes the last step in the biosynthesis of the human Sd(a) antigen through the addition of an N-acetylgalactosamine residue via a beta-1,4 linkage to a subterminal galactose residue substituted with an alpha-2,3-linked sialic acid. B4GALNT2 also catalyzes the last step in the biosynthesis of the Cad antigen (Montiel et al., 2003 [PubMed 12678917]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen B4GALNT2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene B4GALNT2
Gene Accession No. Q09199, Q8NHY0
Gene Alias AI593864; B4galnt2; B4GALT; beta 1,4 N-acetylgalactosaminyltransferase/betal,4 GalNAcT/Sda-GalNAcT; beta-1,4 N-acetylgalactosaminyltransferase 2; beta-1,4-N-acetyl-galactosaminyl transferase 2; beta-1,4-N-acetyl-galactosaminyltransferase 2; beta-4-N-acetylgalactosaminyltransferase; CT-Gal NAc transferase; Dlb1; Dlb-1; dolichos lectin binding 1; Galg2; Galgt2; Ggm3; Igf2bp1; N-acetylgalactosaminyltransferase; sd(a) beta-1,4-GalNAc transferase; sda beta-1,4-GalNAc transferase; UDP-GalNAc:Neu5Aca2-3Galb-R b1,4-N-acetylgalactosaminyltransferase; UDP-GalNAc:Neu5Acalpha2-3Galbeta-R beta1,4-N-acetylgalactosaminyltransferase; UDP-N-acetyl-alpha-D-galactosamine:(N-acetylneuraminyl)-galactosyl-N-acetylglucosaminylpolypeptide-beta-1, 4-N-acetylgalactosaminyltransferase
Gene Symbols B4GALNT2
Host Species Rabbit
Immunogen A synthesized peptide derived from human B4GALNT2(Accession Q8NHY0), corresponding to amino acid residues G37-M87.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124872, 14422
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.