Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B4GALNT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112335
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112335 Supplier Invitrogen™ Supplier No. PA5112335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

B4GALNT3 Polyclonal Antibody for Western Blot

Transfers N-acetylgalactosamine (GalNAc) from UDP-GalNAc to N-acetylglucosamine-beta-benzyl with a beta-1,4-linkage to form N,N'-diacetyllactosediamine, GalNAc-beta-1,4-GlcNAc structures in N-linked glycans and probably O-linked glycans. Mediates the N,N'- diacetyllactosediamine formation on gastric mucosa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B4GALNT3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene B4GALNT3
Gene Accession No. Q6L9W6
Gene Alias B4GalNAcT3; B4GALNT3; beta-1,4-N-acetyl-galactosaminyl transferase 3; beta-1,4-N-acetylgalactosaminyltransferase 3; beta-1,4-N-acetyl-galactosaminyltransferase 3; Beta-1,4-N-acetylgalactosaminyltransferase III; beta4GalNAcT3; Beta4GalNAc-T3; N-acetyl-beta-glucosaminyl-glycoprotein 4-beta-N- acetylgalactosaminyltransferase; N-acetyl-beta-glucosaminyl-glycoprotein 4-beta-N-acetylgalactosaminyltransferase 2; NGalNAc-T2
Gene Symbols B4GALNT3
Host Species Rabbit
Immunogen E. coli-derived Human B4GALNT3 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283358
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.