Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B4GALT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117139
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117139 Supplier Invitrogen™ Supplier No. PA5117139
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: LO2, HeLa, 22Rv1 Immunogen sequence: RDLSRLPQLV GVSTPLQGGS NSAAAIGQSS GELRTGGARP PPPLGASSQP RPGGDSSPVV DSGPGPASNL TSVPVPHTTA LSLPACPEES PLLVGPMLIE FNMPVDLELV AKQNPNVKMG GRYAPRDCVS PHKVAIIIPF RNRQEHLKYW LYYLHPVLQR QQLDYG.

This gene is one of seven beta-1,4-galactosyltransferase (beta4GalT) genes. They encode type II membrane-bound glycoproteins that appear to have exclusive specificity for the donor substrate UDP-galactose; all transfer galactose in a beta1,4 linkage to similar acceptor sugars: GlcNAc, Glc, and Xyl. Each beta4GalT has a distinct function in the biosynthesis of different glycoconjugates and saccharide structures. As type II membrane proteins, they have an N-terminal hydrophobic signal sequence that directs the protein to the Golgi apparatus and which then remains uncleaved to function as a transmembrane anchor. By sequence similarity, the beta4GalTs form four groups: beta4GalT1 and beta4GalT2, beta4GalT3 and beta4GalT4, beta4GalT5 and beta4GalT6, and beta4GalT7. This gene is unique among the beta4GalT genes because it encodes an enzyme that participates both in glycoconjugate and lactose biosynthesis. For the first activity, the enzyme adds galactose to N-acetylglucosamine residues that are either monosaccharides or the nonreducing ends of glycoprotein carbohydrate chains. The second activity is restricted to lactating mammary tissues where the enzyme forms a heterodimer with alpha-lactalbumin to catalyze UDP-galactose + D-glucose <=> UDP + lactose. The two enzymatic forms result from alternate transcription initiation sites and post-translational processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B4GALT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene B4GALT1
Gene Accession No. P15291
Gene Alias b1,4-Galactosyltransferase I; B-1,4-GalT; B-1,4-GalT1; B4galt1; b4Gal-T1; Beta-1,4-galactosyltransferase 1; beta-1,4-GalT; beta-1,4-GalT1; beta-1,4-GalTase 1; beta4Gal-T1; Beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase; Beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase; CDG2D; galactosyltransferase 2 beta 1, 4; GalT; Ggtb; GGTB2; glycoprotein galactosyltransferase beta 1, 4; glycoprotein-4-beta-galactosyltransferase 2; Golgi complex; GT1; GTB; lactose synthase; Lactose synthase A protein; N-acetyllactosamine synthase; Nal synthase; Processed beta-1; Processed beta-1,4-galactosyltransferase 1; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 1; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 1; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 1
Gene Symbols B4GALT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 45-210 of human B4GALT1 (NP_0014882).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2683
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.