Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B4GALT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 203301AP150UL

Description
B4GALT2 is one of type II membrane-bound glycoproteins that appear to have exclusive specificity for the donor substrate UDP-galactose; all transfer galactose in a beta1,4 linkage to similar acceptor sugars: GlcNAc, Glc, and Xyl. Each beta4GalT has a distinct function in the biosynthesis of different glycoconjugates and saccharide structures. As type II membrane proteins, they have an N-terminal hydrophobic signal sequence that directs the protein to the Golgi apparatus and which then remains uncleaved to function as a transmembrane anchor. By sequence similarity, the beta4GalTs form four groups: beta4GalT1 and beta4GalT2, beta4GalT3 and beta4GalT4, beta4GalT5 and beta4GalT6, and beta4GalT7. The enzyme synthesizes N-acetyllactosamine in glycolipids and glycoproteins. Its substrate specificity is affected by alpha-lactalbumin but it is not expressed in lactating mammary tissue.Specifications
B4GALT2 | |
Polyclonal | |
Unconjugated | |
B4GALT2 | |
B4Gal T2, B4Gal T3, B4GALT2, Beta 1,4 GalTase 2, beta4Gal T2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
313536, 53418, 8704 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60909, Q9Z2Y2 | |
B4galt2 | |
B4GALT2 Fusion Protein Ag14046 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction