Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B4GALT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106618
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106618 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106618 Supplier Invitrogen™ Supplier No. PA5106618
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total B4GALT3.

The beta-1,3 galactosyltransferase( beta3GalT) gene family encodes a set of type II transmembrane glycoproteins that are catalytically diverse and use different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine ) to catalyze the addition of an activated monosaccharide to a terminal lactose. The protein coding sequences for beta-1,3-Gal-T genes comprise a single exon and are distantly related to the Drosophila Brainiac gene. The beta-1,4-galactosyltransferase ( beta4GalT) gene family encodes type II membrane-bound glycoproteins that show exclusive specificity for the donor substrate, UDP-galactose. beta-1,4Gal-T genes transfer galactose in a beta-1,4 linkage to similar acceptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B4GALT3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene B4GALT3
Gene Accession No. O60512, Q6P768, Q91YY2
Gene Alias 9530061M23Rik; AA104562; AW125175; B4GALT3; b4Gal-T3; Beta-1,4-galactosyltransferase 3; beta-1,4-galactosyltransferase III; beta-1,4-GalTase 3; beta4Gal-T3; beta4GalT-III; Beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase; beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 3; Beta-N-acetylglucosaminyl-glycolipid4-galactosyltransferase; Beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase; Beta-N-acetylglucosaminylglycopeptide4-galactosyltransferase; ESTM26; ESTM6; N-acetyllactosamine synthase; Nal synthase; R74981; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 3; UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 3; UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 3; beta-1,4-galactosyltransferase III; beta4GalT-III; expressed sequence tag mouse EST 6; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 3; UDP-galactose:beta-N-acetylglucosamine; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 3
Gene Symbols B4GALT3
Host Species Rabbit
Immunogen A synthesized peptide derived from human B4GALT3(Accession O60512), corresponding to amino acid residues L271-S321.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 494342, 57370, 8703
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.