Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B4GALT3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539058
Description
The beta-1,3 galactosyltransferase( beta3GalT) gene family encodes a set of type II transmembrane glycoproteins that are catalytically diverse and use different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine ) to catalyze the addition of an activated monosaccharide to a terminal lactose. The protein coding sequences for beta-1,3-Gal-T genes comprise a single exon and are distantly related to the Drosophila Brainiac gene. The beta-1,4-galactosyltransferase ( beta4GalT) gene family encodes type II membrane-bound glycoproteins that show exclusive specificity for the donor substrate, UDP-galactose. beta-1,4Gal-T genes transfer galactose in a beta-1,4 linkage to similar acceptor.
Specifications
| B4GALT3 | |
| Polyclonal | |
| Unconjugated | |
| B4GALT3 | |
| Beta-1,4-galactosyltransferase 3, Beta-1,4-GalTase 3, Beta4Gal-T3, b4Gal-T3, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 3, UDP-galactose:beta-N-acetylglucosamine, beta-1,4-galactosyltransferase 3, N-acetyllactosamine synthase, Nal synthase, Beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase, Beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase, B4GALT3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 494342, 57370, 8703 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60512, Q91YY2, Q6P768 | |
| B4GALT3 | |
| A synthetic peptide derived from the internal region of human B4GALT3 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction