Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B4GALT4 Antibody (OTI8B6), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271385UV
Description
B4GALT4 Monoclonal specifically detects B4GALT4 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
B4GALT4 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
B4GALT4 | |
Recombinant protein fragment corresponding to amino acids 40-322 of human B4GALT4 (NP_003769) produced in E.coli. | |
0.1 mL | |
8702.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI8B6 | |
Western Blot, Immunohistochemistry | |
B4Gal-T4, beta-1,4-galactosyltransferase 4, Beta-1,4-GalTase 4, beta4Gal-T4, beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 4, EC 2.4.1.-, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase 4, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 4, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 4, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 4 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction