Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ B4GALT4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561342
Description
Immunogen sequence: AIQEIPKAKE FMANFHKTLI LGKGKTLTNE ASTKKVELDN CPSVSPYLRG QSKLIFKPDL TLEEVQAENP KVSRGRYRPQ ECKALQRVAI LVPHRNREKH LMYLLEHLHP F Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 77%.
This gene is one of seven beta-1,4-galactosyltransferase (beta4GalT) genes. They encode type II membrane-bound glycoproteins that appear to have exclusive specificity for the donor substrate UDP-galactose; all transfer galactose in a beta1,4 linkage to similar acceptor sugars: GlcNAc, Glc, and Xyl. Each beta4GalT has a distinct function in the biosynthesis of different glycoconjugates and saccharide structures. As type II membrane proteins, they have an N-terminal hydrophobic signal sequence that directs the protein to the Golgi apparatus and which then remains uncleaved to function as a transmembrane anchor. By sequence similarity, the beta4GalTs form four groups: beta4GalT1 and beta4GalT2, beta4GalT3 and beta4GalT4, beta4GalT5 and beta4GalT6, and beta4GalT7. The enzyme encoded by this gene appears to mainly play a role in glycolipid biosynthesis. Two alternatively spliced transcript variants have been found for this gene.
Specifications
B4GALT4 | |
Polyclonal | |
Unconjugated | |
B4GALT4 | |
9130402O08Rik; B4GALT4; b4Gal-T4; B4galt-IV; Beta-1,4-galactosyltransferase 4; beta-1,4-galactosyltransferase IV; beta-1,4-GalTase 4; beta4Gal-T4; beta4GalT-IV; beta-N-acetylglucosaminyl; Beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase; beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 4; Lactotriaosylceramide beta-1,4-galactosyltransferase; N-acetyllactosamine synthase; Nal synthase; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase 4; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 4; UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 4; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 4; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 4; UNQ552/PRO1109 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8702 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60513 | |
B4GALT4 | |
Recombinant protein corresponding to Human B4GALT4. Recombinant protein control fragment (Product #RP-90163). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction