Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B4GALT4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7247040 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166126 20 μg
NB166125 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166126 Supplier Novus Biologicals Supplier No. NBP31551520UL
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

B4GALT4 Polyclonal antibody specifically detects B4GALT4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen B4GALT4
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200
Formulation PBS, 50% glycerol, pH7.3
Gene Alias B4Gal-T4, beta-1,4-galactosyltransferase 4, Beta-1,4-GalTase 4, beta4Gal-T4, beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 4, EC 2.4.1.-, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase 4, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 4, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 4, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 35-344 of human B4GALT4 (NP_003769.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8702
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.