Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B4GALT5 Antibody, Novus Biologicals™
SDP

Catalog No. p-7108651 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP169018 100 μL
NBP16901820 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP169018 Supplier Novus Biologicals Supplier No. NBP169018
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

B4GALT5 Polyclonal specifically detects B4GALT5 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen B4GALT5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9JMK0
Gene Alias B4Gal-T5, beta-1,4-galactosyltransferase 5, Beta-1,4-GalT II, beta-1,4-GalT IV, Beta-1,4-GalTase 5, beta-1.4-galactosyltransferase V, beta4-GalT IV, Beta4Gal-T5, BETA4-GALT-IV, beta4GalT-V, EC 2.4.1.-, gt-V, MGC138470, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 5, UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 5, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 5, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 5
Gene Symbols B4GALT5
Host Species Rabbit
Immunogen Synthetic peptides corresponding to B4galt5 (UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 5) The peptide sequence was selected from the C terminal of B4galt5. Peptide sequence GYSVSRPEGDTGKYKSIPHHHRGEVQFLGRYALLRKSKERQGLDGLNNLN The peptide sequence for this immunogen was taken from within the described region.
Molecular Weight of Antigen 45 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9334
Test Specificity Zebrafish: 85%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.