Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B4GALT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112886
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112886 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112886 Supplier Invitrogen™ Supplier No. PA5112886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

B4GALT5 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene is one of seven beta-1,4-galactosyltransferase (beta4GalT) genes. They encode type II membrane-bound glycoproteins that appear to have exclusive specificity for the donor substrate UDP-galactose; all transfer galactose in a beta1,4 linkage to similar acceptor sugars: GlcNAc, Glc, and Xyl. Each beta4GalT has a distinct function in the biosynthesis of different glycoconjugates and saccharide structures. As type II membrane proteins, they have an N-terminal hydrophobic signal sequence that directs the protein to the Golgi apparatus and which then remains uncleaved to function as a transmembrane anchor. By sequence similarity, the beta4GalTs form four groups: beta4GalT1 and beta4GalT2, beta4GalT3 and beta4GalT4, beta4GalT5 and beta4GalT6, and beta4GalT7. The function of the enzyme encoded by this gene is not clear. This gene was previously designated as B4GALT4 but was renamed to B4GALT5. In the literature it is also referred to as beta4GalT2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B4GALT5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene B4GALT5
Gene Accession No. O43286, Q9JMK0
Gene Alias 4-gal; 9430078I07Rik; AW049941; AW539721; B4GALT5; B4Gal-T5; Beta-1,4-galactosyltransferase 5; beta-1,4-galactosyltransferase V; beta-1,4-GalT II; beta-1,4-GalT IV; beta-1,4-GalTase 5; beta-1.4-galactosyltransferase V; beta4-GalT IV; beta4Gal-T5; BETA4-GALT-IV; beta4GalT-V; Bgt-5; Glucosylceramide beta-1,4-galactosyltransferase; gt-V; LacCer synthase; Lactosylceramide synthase; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 5; UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 5; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 5; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 5
Gene Symbols B4GALT5
Host Species Rabbit
Immunogen Recombinant Human Beta-1,4-galactosyltransferase 5 protein (101-388AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56336, 9334
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.