Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B4GALT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563547
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563547 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563547 Supplier Invitrogen™ Supplier No. PA563547
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTYLPENFTY SPYLPCPEKL PYMRGFLNVN VSEVSFDEIH QLFSKDLDIE PGGHWRPKDC KPRWK Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.

This gene is one of seven beta-1,4-galactosyltransferase (beta4GalT) genes. They encode type II membrane-bound glycoproteins that appear to have exclusive specificity for the donor substrate UDP-galactose; all transfer galactose in a beta1,4 linkage to similar acceptor sugars: GlcNAc, Glc, and Xyl. Each beta4GalT has a distinct function in the biosynthesis of different glycoconjugates and saccharide structures. As type II membrane proteins, they have an N-terminal hydrophobic signal sequence that directs the protein to the Golgi apparatus and which then remains uncleaved to function as a transmembrane anchor. By sequence similarity, the beta4GalTs form four groups: beta4GalT1 and beta4GalT2, beta4GalT3 and beta4GalT4, beta4GalT5 and beta4GalT6, and beta4GalT7. The enzyme encoded by this gene is a lactosylceramide synthase important for glycolipid biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B4GALT6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene B4GALT6
Gene Accession No. Q9UBX8
Gene Alias AA536803; AU022389; B4galt6; B4Gal-T6; beta-1, 4-galactosyltransferase VI; beta-1,4-galactosyltransferase 6; Beta-1,4-GalTase 6; beta4Gal-T6; beta4GalT-VI; GalT; Glucosylceramide beta-1,4-galactosyltransferase; LacCer synthase; Lactosylceramide synthase; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 6; UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 6; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 6; UDP-Gal:glucosylceramide beta-1,4-galactosyltransferase; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 6; zgc:55730
Gene Symbols B4GALT6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human B4GALT6. Recombinant protein control fragment (Product #RP-103322).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9331
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.