Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ B4GALT7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568114
Description
Predicted to react with Mouse and Rat samples.
Required for the biosynthesis of the tetrasaccharide linkage region of proteoglycans, especially for small proteoglycans in skin fibroblasts.
Specifications
| B4GALT7 | |
| Polyclonal | |
| Unconjugated | |
| B4GALT7 | |
| B4galt7; b4Gal-T7; Beta-1,4-galactosyltransferase 7; beta-1,4-galactosyltransferase VII; beta-1,4-GalTase 7; beta4Gal-T7; beta4GalT-VII; EDSP1; EDSSLA; galactosyltransferase I; proteoglycan UDP-galactose:beta-xylose beta1,4-galactosyltransferase I; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 7; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 7; UDP-galactose:beta-xylose beta-1,4-galactosyltransferase; UNQ748/PRO1478; XGALT1; XGalT-1; XGPT; XGPT1; Xylosylprotein 4-beta-galactosyltransferase; xylosylprotein beta 1,4-galactosyltransferase, polypeptide 7; Xylosylprotein beta-1,4-galactosyltransferase; xylosylprotein beta1,4-galactosyltransferase, polypeptide 7 (galactosyltransferase I) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11285, 218271, 364675 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8R087, Q9UBV7 | |
| B4GALT7 | |
| Synthesized peptide derived from human B4GALT7. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction