Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-1/CD80 Antibody (018), HRP

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290591H
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Mouse samples. It is validated for ELISA.Specifications
B7-1/CD80 | |
Monoclonal | |
HRP | |
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse B7-1/CD80 (Q00609-1; Met1-Lys245). | |
0.1 mL | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction | |
941 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
18 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction